The Israel Medical Association journal : IMAJ
Association between Enlarged Axillary Lymph Nodes and Silicone Breast Implant Ruptures seen on Magnetic Resonance Imaging.
PMID 28457073
Silicone breast augmentation is a common cosmetic surgery. Previous case reports demonstrated lymphadenopathy in the presence of implant ruptures. To investigate the association between enlarged axillary lymph nodes and silicone implant ruptures as seen on breast magnetic resonance imaging (MRI). Two groups were derived retrospectively from breast MRI reports in our institution for the period December 2011-May 2014. A search of our hospital records for "silicone" and "lymph node" was performed (group A), and the relationship between the presence of enlarged nodes and ruptures was evaluated. The prevalence of ruptures in the presence of nodes was calculated and the association between MRI imaging features and ruptures evaluated. A search for "silicone" and "implant rupture" was performed (group B) and, as for group A, the relationship between the presence of ruptures and nodes was evaluated and the prevalence of enlarged nodes in the presence of ruptures calculated. Group A comprised 45 women with enlarged nodes. Intracapsular ruptures were associated with nodes (P = 0.005), while extracapsular ruptures showed a trend of association with nodes (P = 0.08). The prevalence of ruptures in the presence of nodes was 31.4%. Nodes associated with ruptures showed a strong silicone signal (P = 0.008) and absent enhancement (P = 0.005). Group B comprised 73 women with ruptures. Enlarged nodes were associated with both intra- and extracapsular ruptures (P < 0.001 and P = 0.002 respectively). The prevalence of nodes in the presence of ruptures was 22.2%. Enlarged axillary nodes were associated with ruptures in two groups of patients. This finding can guide clinical decisions when either enlarged nodes or ruptures are encountered in patients with silicone implants. The association between silicone lymphadenopathy and implant rupture raises concerns regarding the role of rupture in silicone-induced systemic disease.
Related Materials
Product #
Molecular Formula
Add to Cart
L-Aspartic acid β-hydroxamate, serine racemase inhibitor