EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Design, synthesis, and identification of a novel napthalamide-isoselenocyanate compound NISC-6 as a dual Topoisomerase-IIα and Akt pathway inhibitor, and evaluation of its anti-melanoma activity.
PMID 28458134
Abstract
Synthesis and anti-melanoma activity of novel naphthalimide isoselenocyanate (NISC) and naphthalimide selenourea (NSU) analogs are described. The novel agents were screened for growth inhibition of different human melanoma cell lines including those having BRAF