EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
Functional β
PMID 28459158
Abstract
Altered sensitivity to the chronotropic effect of catecholamines and a reduction in the β
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7971
Cilostamide, phosphodiesterase inhibitor
C20H26N2O3