EMAIL THIS PAGE TO A FRIEND
SLAS discovery : advancing life sciences R & D
Identification of Human UMP/CMP Kinase 1 as Doxorubicin Binding Target Using Protein Microarray.
PMID 28459633
Abstract
Doxorubicin (DOX) is a leading anthracycline drug with exceptional efficacy; however, little is known about the molecular mechanisms of its side effects, which include heart muscle damage, noncancerous cell death, and drug resistance. A total of 17,950 human proteins expressed in HEK293 cells were screened and yielded 14 hits. Competitive and binding experiments further verified the binding of DOX to UMP/CMP kinase 1 (CMPK1), and microscale thermophoresis showed that DOX binds to CMPK1 with a K
Related Materials