EMAIL THIS PAGE TO A FRIEND
PLoS neglected tropical diseases
The clinical features of 590 patients with brucellosis in Xinjiang, China with the emphasis on the treatment of complications.
PMID 28459811
Abstract
This study aims to analyze the clinical characteristics and treatment outcomes of 590 patients with brucellosis in Xinjiang, China. The clinical characteristics, laboratory findings, complications and prognosis of 590 patients infected with brucellosis were retrospectively analyzed. These patients had a mean age of 44.24 ± 15.83 years with 60.5% having a history of close contacting with cattle and sheep. Of them, 53.6% (316 /590) were in acute phase and 21.5% were in chronic phase. Agglutination test showed 98.5% positive with 34% blood culture positive of Brucella. The major symptoms were fatigue (91%), hyperhidrosis(88.1%), fever(86.9%), and joint pain(81%) with 29.8% having enlarged liver, 26.1% having enlarged spleen and 23.2% having osteoarticular complications. Combination of doxycycline plus rifampicin for 12 weeks was an effective regimen for patients without complications. The 3-drug regimen (doxycycline+rifampicin+levofloxacin) for 12 weeks was recommended for these with complications. There were 6 patients died (1.02%) with overall relapse rate of 5.98%. Brucellosis is mostly associated with contacting with domestic animal production in Xinjiang, China. Clinical symptoms include fever, fatigue, hyperhidrosis, and joint pain with common complication of osteoarticular involvement. Three-drug-regimen of doxycycline+rifampicin+levofloxacin for 12 weeks was effective for these patients with complications.