EMAIL THIS PAGE TO A FRIEND
Canadian journal of physiology and pharmacology
cAMP attenuates angiotensin-II-induced Egr-1 expression via PKA-dependent signaling pathway in vascular smooth muscle cells.
PMID 28460186
Abstract
cAMP has been shown to inhibit vascular smooth muscle cell proliferation and exerts a vasculoprotective effect. An upregulation of the early growth response protein-1 (Egr-1) expression has been linked with the development of atherosclerosis and intimal hyperplasia. We have recently demonstrated that angiotensin-II (Ang-II) stimulates Egr-1 expression via Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9525
Angiotensin II human, ≥93% (HPLC), powder
C50H71N13O12
586846
N-Benzoyladenosine, 96%
C17H17N5O5