EMAIL THIS PAGE TO A FRIEND
Aquatic toxicology (Amsterdam, Netherlands)
Acute toxicity and esterase response to carbaryl exposure in two different populations of amphipods Hyalella curvispina.
PMID 28460306
Abstract
During the last years, a carbaryl insecticide was extensively applied in the valley of Río Negro and Neuquén, North Patagonia Argentina, to manage codling moths (Cydia pomonella), the main pest of pear and apple trees. In this study carbaryl susceptibility and B-esterase activity from both insecticide-exposed and non-exposed field populations of amphipods Hyalella curvispina were studied. Two subpopulations, one susceptible to carbaryl (LC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8505
1-Naphthyl acetate, ≥98% (C)
C12H10O2
F8761
Fast Garnet GBC sulfate salt, diazonium dye
C14H14N4O4S