EMAIL THIS PAGE TO A FRIEND
Journal of research in medical sciences : the official journal of Isfahan University of Medical Sciences
Assessment of the role of plasma nitric oxide levels, T-786C genetic polymorphism, and gene expression levels of endothelial nitric oxide synthase in the development of coronary artery disease.
PMID 28461820
Abstract
Reduced bioavailability of nitric oxide (NO) and the T-786C polymorphism of endothelial nitric oxide synthase (eNOS) gene have been reported as risk factors for the development of coronary artery disease (CAD) with conflicting results. We investigated the association of plasma NO levels, T-786C genetic polymorphism, and gene expression levels of eNOS with CAD risk in an Iranian subpopulation. Studied population included 100 patients with angiographically verified CAD and 100 ethnically matched controls. Analysis of T-786C genetic polymorphism and gene expression levels of eNOS was conducted by polymerase chain reaction (PCR) restriction fragment length polymorphism and real-time reverse transcription-PCR methods, respectively. Plasma levels of NO were measured using Griess method. The CC genotype distribution (15% vs. 6%, Our study indicated that reduced NO levels and eNOS T-786C genetic polymorphism are significant risk factors for the development and severity of CAD in the Iranian population.