EMAIL THIS PAGE TO A FRIEND
Cell biology international
Altered expression of eNOS, prostacyclin synthase, prostaglandin G/H synthase, and thromboxane synthase in porcine aortic endothelial cells after exposure to human serum-relevance to xenotransplantation.
PMID 28462511
Abstract
Under normal conditions, the activity of platelets is stringently and precisely balanced between activation and quiescent state. This guarantees rapid hemostasis and avoids uncontrolled thrombosis. However, excessive platelet activation and resulting thrombotic microangiopathy are frequently observed in pig-to-primate xenotransplantation models. Endothelium-derived inhibitory mechanisms play an important role in regulation of platelet activation. These mainly include nitric oxide (NO), prostacyclin PGI