EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Physiological functions of pyruvate:NADP
PMID 28463550
Abstract
In Euglena gracilis, pyruvate:NADP
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
63423
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
C12H22O11