EMAIL THIS PAGE TO A FRIEND
Journal of neurotrauma
Mild Traumatic Brain Injury Leads to Decreased Inhibition and a Differential Response of Calretinin Positive Interneurons in the Injured Cortex.
PMID 28463587
Abstract
It is clear that even mild forms of traumatic brain injury (TBI) can have lasting cognitive effects; however, the specific cellular changes responsible for the functional deficits remain poorly understood. Previous studies suggest that not all neurons respond in the same way and that changes to neuronal architecture may be subtype specific. The current study aimed to characterize the response of interneurons to TBI. To model TBI in vitro, the neurites of primary cortical neurons were transected at 15 days in vitro. In response, calretinin