EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Crown Ether Complexes of Alkali-Metal Chlorides from SO
PMID 28464436
Abstract
The structures of alkali-metal chloride SO
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
194905
12-Crown-4, 98%
C8H16O4