EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
A liquid chromatography-tandem mass spectrometry method to measure fatty acids in biological samples.
PMID 28467947
Abstract
As pre-clinical and clinical research interest in ω-3 polyunsaturated fatty acids (PUFA) increases, so does the need for a fast, accurate and reproducible analytical method to measure fatty acids (FA) in biological samples in order to validate potential prognostic and predictive biomarkers, as well as establishing compliance in ω-3 PUFA intervention trials. We developed a LC-ESI-MS/MS method suitable for high throughput development to measure FAs and validated it in the context of treatment with the ω-3 PUFA eicosapentaenoic acid (EPA). Uniquely we directly compared the LC-ESI-MS/MS method to a GC-MS protocol. We demonstrated the LC-ESI-MS/MS method is accurate and reproducible, with coefficients of variation consistently below 15% for each PUFA analysed. The relative FA content values correlated well with those obtained by GC-MS (r