EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Cell physiology
ENaC activity is regulated by calpain-2 proteolysis of MARCKS proteins.
PMID 28468944
Abstract
We previously demonstrated a role for the myristoylated alanine-rich C kinase substrate (MARCKS) to serve as an adaptor protein in the anionic phospholipid phosphate-dependent regulation of the epithelial sodium channel (ENaC). Both MARCKS and ENaC are regulated by proteolysis. Calpains are a family of ubiquitously expressed intracellular Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SCP0063 Calpastatin Peptide Ac 184-210, ≥95% (HPLC)
C142H230N36O44S1