EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Renal physiology
Luminal ANG II is internalized as a complex with AT
PMID 28468964
Abstract
ANG II has many biological effects in renal physiology, particularly in Ca