EMAIL THIS PAGE TO A FRIEND
Biotechnology for biofuels
Characterisation of non-degraded oligosaccharides in enzymatically hydrolysed and fermented, dilute ammonia-pretreated corn stover for ethanol production.
PMID 28469707
Abstract
Corn stover is lignocellulosic biomass that has potential to be used as raw material for bioethanol production. In the current research, dilute ammonia pretreatment was used to improve the accessibility of corn stover carbohydrates to subsequently added hydrolytic enzymes. Some carbohydrates, however, were still present after enzymatic hydrolysis and fermentation. Hence, this research was aimed to characterise the recalcitrant carbohydrates, especially the oligosaccharides that remained after hydrolysis and fermentation of dilute ammonia-pretreated corn stover (DACS). About 35% (w/w) of DACS carbohydrates remained after enzymatic hydrolysis and fermentation of the released monosaccharides. One-third of these recalcitrant carbohydrates were water soluble and composed of diverse oligosaccharides. By using UHPLC-MS Around one-third of the 35% (w/w) recalcitrant DACS carbohydrates remained as water-soluble saccharides. In this study, more than 50 recalcitrant oligosaccharides were detected, which mostly composed of xylosyl and/or glucosyl residues. The most pronounced oligosaccharides were UAXOS and XGOS. Hence, α-glucuronidase and α-xylosidase were suggested to be added to the enzyme mixture to degrade these oligosaccharides further, and hence the fermentation yield is potentially increased.