EMAIL THIS PAGE TO A FRIEND
Frontiers in cellular and infection microbiology
Signaling Mediated by Toll-
PMID 28469996
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU155971 MISSION® esiRNA, esiRNA human TLR5 (esiRNA1)