EMAIL THIS PAGE TO A FRIEND
Alimentary pharmacology & therapeutics
Concentration-dependent response to pioglitazone in nonalcoholic steatohepatitis.
PMID 28470881
Abstract
Pioglitazone is a safe and effective option to manage patients with type 2 diabetes and nonalcoholic steatohepatitis (NASH). However, there is marked variability in treatment response. To evaluate the relationship between concentrations of pioglitazone and its active metabolites and treatment outcomes in patients with NASH. Pioglitazone concentrations were measured in patients with NASH treated with pioglitazone 45 mg/day for 18 months; liver biopsy samples were obtained at baseline and after treatment. The primary outcome was a ≥2-point reduction in NAFLD activity score (NAS) with at least one-point improvement in more than one liver histology category and without worsening of fibrosis. A novel marker, the pioglitazone exposure index, was calculated to consider the concentrations of pioglitazone as well as the two active metabolites. The response to pioglitazone was concentration-dependent as evidenced by the significant relationship between both pioglitazone concentration and pioglitazone exposure index with changes in NAS (r=.48, P=.0002 and r=.51, P<.0001, respectively), steatosis (r=.41, P=.002 and r=.46, P=.0005), and inflammation (r=.44, P=.0009 and r=.40, P=.0003). The pioglitazone exposure index was also associated with a change in ballooning (P=.04). The pioglitazone exposure index was higher in patients with NASH resolution (2.85±1.38 vs 1.78±1.48, P=.018). A predictive model for the primary outcome was developed that incorporated baseline NAS and pioglitazone exposure index (AUC=0.77). This study demonstrates the importance of pioglitazone exposure to variable response in patients with NASH, and indicates potential factors that may identify patients most likely to benefit from chronic pioglitazone treatment.