EMAIL THIS PAGE TO A FRIEND
Tissue engineering. Part A
Interleukin-10 Deficiency Impairs Reparative Properties of Bone Marrow-Derived Endothelial Progenitor Cell Exosomes.
PMID 28471299
Abstract
Endothelial progenitor cell (EPC)-based therapy has immense potential to promote cardiac neovascularization and attenuate ischemic injury. Functional benefits of EPCs and other adult stem cell therapies largely involve paracrine mechanisms and exosomes secreted by stem cells are emerging as pivotal paracrine entity of stem/progenitor cells. However, modest outcomes after EPC-/stem cell-based clinical trials suggest that stem cell/exosome function might be modulated by stimuli they encounter in ischemic tissues, including systemic inflammation. We hypothesized that EPCs under inflammatory stress might produce exosomes of altered and dysfunctional content, which may compromise EPC repair in ischemic heart disease. We have previously shown that EPCs obtained from interleukin-10 knockout (IL-10KO) mice (model mimicking systemic inflammation) display impaired angiogenic functions. Whether IL-10KO-EPC-derived exosomes inherit their parental dysfunctional phenotype and whether inflammatory environment alters the cargo of their secreted exosomes are not known. After cell expansion from IL-10KO and wild-type (WT) mice, we isolated exosomes and compared their functions in terms of effect on cell survival, proliferation, migration, and angiogenic capacity in vitro. WT-EPC-Exo treatment enhanced endothelial cell proliferation and tube formation, and inhibited apoptosis, whereas IL-10KO-Exo exhibited impaired or even detrimental effects, suggesting that the reparative capacity of WT-EPC-Exo is lost in exosomes derived from IL-10-KO-EPCs. Deep RNA sequencing and proteomic analyses to compare WT and IL-10KO-Exo revealed drastically altered exosome cargo. Importantly, IL-10KO-EPC-Exo were highly enriched in microRNAs and proteins that promote inflammation and apoptosis and inhibit angiogenesis. Through modulation of a specific enriched miRNA (miR-375), we partially rescued IL-10KO-EPC-Exo dysfunction. Thus, our study revealed that EPC exosomes display impaired function under inflammatory stimulus through changed exosome contents, and the dysfunction can be rescued by modulation of a specific target packed in exosomes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU016511 MISSION® esiRNA, esiRNA targeting human CHN1 (esiRNA1)