EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Lung cellular and molecular physiology
Therapeutic blockade of CD54 attenuates pulmonary barrier damage in T cell-induced acute lung injury.
PMID 28473322
Abstract
Acute respiratory distress syndrome (ARDS) is a serious, often fatal condition without available pharmacotherapy. Although the role of innate cells in ARDS has been studied extensively, emerging evidence suggests that T cells may be involved in disease etiology.
Related Materials