EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
TCTP regulates spindle assembly during postovulatory aging and prevents deterioration in mouse oocyte quality.
PMID 28476647
Abstract
If no fertilization occurs for a prolonged time following ovulation, oocytes experience a time-dependent deterioration in quality both in vivo and in vitro due to processes called postovulatory aging. Because the postovulatory aging of oocytes has marked detrimental effects on embryo development and offspring, many efforts have been made to unveil the underlying mechanisms. Here we showed that translationally controlled tumor protein (TCTP) regulates spindle assembly during postovulatory aging and prevents deterioration in mouse oocyte quality. Spindle dynamics decreased with reduced TCTP level during aging of mouse oocytes. Knockdown of TCTP accelerated the reduction of spindle dynamics, accompanying with aging-related deterioration of oocyte quality. Conversely, overexpression of TCTP prevented aging-associated decline of spindle dynamics. Moreover, the aging-related abnormalities in oocytes were rescued after TCTP overexpression, thereby improving fertilization competency and subsequent embryo development. Therefore, our results demonstrate that TCTP-mediated spindle dynamics play a key role in maintaining oocyte quality during postovulatory aging and overexpression of TCTP is sufficient to prevent aging-associated abnormalities in mouse oocytes.