EMAIL THIS PAGE TO A FRIEND
Anticancer research
Prediction of Relapse After Therapy Withdrawal in Women with Endometrial Hyperplasia: A Long-term Follow-up Study.
PMID 28476823
Abstract
To investigate whether risk of relapse of endometrial hyperplasia persists many years after successful primary therapy and whether clinical or biological markers observed at primary diagnosis may predict relapse. A series of 57 women with endometrial hyperplasia received levonorgestrel-impregnated intrauterine system or oral progestin for three months during 1998-2000. Index biopsies were classified according to WHO1994 and D-score systems, and immunohistochemical staining for estrogen receptor α (ERα), estrogen receptor β (ERβ), progesterone receptor A (PRA), progesterone receptor B (PRB), B-cell lymphoma 2/apoptosis regulator (BCL2), BCL2-associated X protein/apoptosis regulator (BAX), paired box 2 (PAX2), and phosphatase and tensin homolog (PTEN) reported as H-scores. Over a follow-up of 157.8 months, 23% (10/43) of patients experienced relapse. No correlation with age, body mass index, parity, WHO94 classification, or D-score was found. Only PRA (p=0.004) and PRB (p=0.038) showed certain correlation with relapse. Endometrial hyperplasia recurs many years after successful progestin therapy. Increased expression of PRB and reduced expression of PRA significantly correlated with relapse. Our results support the importance of continuous endometrial protection and the need for new clinical surveillance guidelines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
20357
ORBO 2-HMP on Supelpak 20N Tube, W,W,W separators, O.D. × L 8 mm × 115 mm, pkg of 25 ea
20257-U
ORBO 23 2-HMP on Amberlite® XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
20231
ORBO 24 2-HMP on Amberlite® XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
20832-U ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20034 ORBO 403 Tenax® TA specially treated (60/80), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 50 ea
20262 ORBO 42 Small Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 66/33 mg, W,F,F (specially treated) separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20258 ORBO 43 Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20260-U ORBO 44Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
20350
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite® XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
20030-U
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20032
ORBO 506 Activated Silica Gel (45/60) 300/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20263
ORBO 52 Large Activated Silica Gel (45/60) 150/150 mg, W,W,W separators, O.D. × L 8 mm × 75 mm, pkg of 50 ea
20229
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20265
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
20033
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20049 ORBO 605 Amberlite® XAD®-2 specially cleaned (20/50) 100/50 mg, W,W,W separators, O.D. × L 6 mm × 100 mm, pkg of 50 ea
20050-U ORBO 608 Amberlite® XAD®-2 (20/50) 150/75 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20051 ORBO 609 Amberlite® XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
20052 ORBO 613 Amberlite® XAD®-4 80/40 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
20029-U ORBO 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
20812-U ORBO 80 Coated Filter, with cassettes (unassembled)
20811
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
20069
ORBO 827 LpDNPH Coated Filter, 37mm, pkg of 25 ea
20362 ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate