EMAIL THIS PAGE TO A FRIEND
Journal of biomolecular NMR
Comparison of the free and ligand-bound imino hydrogen exchange rates for the cocaine-binding aptamer.
PMID 28477231
Abstract
Using NMR magnetization transfer experiments, the hydrogen exchange rate constants (k