EMAIL THIS PAGE TO A FRIEND
Plant, cell & environment
Arabidopsis ketoacyl-CoA synthase 16 (KCS16) forms C
PMID 28477442
Abstract
The aliphatic waxes sealing plant surfaces against environmental stress are generated by fatty acid elongase complexes, each containing a β-ketoacyl-CoA synthase (KCS) enzyme that catalyses a crucial condensation forming a new C─C bond to extend the carbon backbone. The relatively high abundance of C