EMAIL THIS PAGE TO A FRIEND
Environmental pollution (Barking, Essex : 1987)
The use of a 0.20 μm particulate matter filter decreases cytotoxicity in lung epithelial cells following air-liquid interface exposure to motorcycle exhaust.
PMID 28477553
Abstract
This study was designed to investigate whether the use of a 0.20 μm particulate matter (PM) filter reduced the cytotoxicity induced by motorcycle exhaust (ME), a mixture of gases and particles, in lung epithelial cells cultured in air-liquid interface (ALI) inserts. The concentrations of PM, carbon monoxide, carbon dioxide, total hydrocarbons (THC), total volatile organic compounds, and nitrogen oxides in both filtered ME (fME) by a 0.20 μm filter and non-filtered ME (non-fME) were measured. Lung epithelial cells were exposed to clean air, fME, or non-fME in the ALI chamber. Cell relative viabilities (CRV) and the reactive oxygen species (ROS) generation were determined. Our results revealed that PM