EMAIL THIS PAGE TO A FRIEND
Journal of the neurological sciences
Usefulness of combining
PMID 28477690
Abstract
Although single-photon emission computerized tomography of the dopamine transporter (DAT-SPECT) is useful for diagnosing parkinsonian syndrome, its applicability toward the early phase of Parkinson's disease remains unknown. We enrolled 32 patients showing parkinsonism with normal cardiac The striatal asymmetry indexes were significantly lower in group 1 than in group 2 (p<0.05), but there were no differences in the specific binding ratios between the two groups. The combined use of striatal asymmetry index on DAT-SPECT and cardiac MIBG scintigraphy might offer useful clues for the differential diagnosis of the early phase Parkinson's disease from other parkinsonian syndromes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69480
N-Methyl-N-trimethylsilylacetamide, for GC derivatization, ≥97.0% (GC)
C6H15NOSi