EMAIL THIS PAGE TO A FRIEND
Molecular and cellular neurosciences
Retinoic acid increases glucocorticoid receptor phosphorylation via cyclin-dependent kinase 5.
PMID 28477983
Abstract
Glucocorticoid receptor (GR) function is modulated by phosphorylation. As retinoic acid (RA) can activate some cytoplasmic kinases able to phosphorylate GR, we investigated whether RA could modulate GR phosphorylation in neuronal cells in a context of long-term glucocorticoid exposure. A 4-day treatment of dexamethasone (Dex) plus RA, showed that RA potentiated the (Dex)-induced phosphorylation on GR Serine 220 (