EMAIL THIS PAGE TO A FRIEND
Journal of the American Oil Chemists' Society
Effect of Fatty Acid Unsaturation on Phytosteryl Ester Degradation.
PMID 28479606
Abstract
This study examined the thermo-oxidative degradation of stigmasterol fatty acids esters. Stigmasterol stearate, oleate, linoleate and linolenate were synthesized by chemical esterification and their purity evaluated by
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C79409
Cholesteryl stearate, 96%
C45H80O2
L2376
Linolenic acid, ≥99%
C18H30O2