EMAIL THIS PAGE TO A FRIEND
Ecology and evolution
Plasticity results in delayed breeding in a long-distant migrant seabird.
PMID 28480009
Abstract
A major question for conservationists and evolutionary biologists is whether natural populations can adapt to rapid environmental change through micro-evolution or phenotypic plasticity. Making use of 17 years of data from a colony of a long-distant migratory seabird, the common tern (