EMAIL THIS PAGE TO A FRIEND
Diabetologia
Retinopathy with central oedema in an INS
PMID 28480495
Abstract
Diabetic retinopathy is a severe complication of diabetes mellitus that often leads to blindness. Because the pathophysiology of diabetic retinopathy is not fully understood and novel therapeutic interventions require testing, there is a need for reliable animal models that mimic all the complications of diabetic retinopathy. Pig eyes share important anatomical and physiological similarities with human eyes. Previous studies have demonstrated that INS Eyes from six INS INS The INS