EMAIL THIS PAGE TO A FRIEND
eLife
Multiple short windows of calcium-dependent protein kinase 4 activity coordinate distinct cell cycle events during
PMID 28481199
Abstract
Malaria transmission relies on the production of gametes following ingestion by a mosquito. Here, we show that Ca