EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
New cycloartane type-triterpenoids from the areal parts of Caragana sukiensis and their biological activities.
PMID 28482219
Abstract
A comprehensive re-investigation of aerial parts of Caragana sukiensis resulted in the isolation of twelve compounds (1-12) including three new cycloartane type triterpenoids (3-5) respectively. Chemical structures of the isolated compounds were established by analysis of their IR, HRMSESI, 1D and 2D NMR spectroscopic data. In addition, these compounds were evaluated for their cytotoxic activity against cancer lines (HeLa, A549, MCF-7, DU-145) and Human embryonic kidney cell line (HEK-293). The results indicated that compound 8 showed potent cytotoxic activity against A549 with IC