EMAIL THIS PAGE TO A FRIEND
Fish & shellfish immunology
Inhibition of Cyclosporine A or rapamycin on T lymphocyte counts and the influence on the immune responses of B lymphocytes in flounder (Paralichthys olivaceus).
PMID 28483552
Abstract
In acquired immunity, T lymphocytes regulate the immune responses of B lymphocytes, including the IgM