EMAIL THIS PAGE TO A FRIEND
Translational psychiatry
Pdxdc1 modulates prepulse inhibition of acoustic startle in the mouse.
PMID 28485732
Abstract
Current antipsychotic medications used to treat schizophrenia all target the dopamine D2 receptor. Although these drugs have serious side effects and limited efficacy, no novel molecular targets for schizophrenia treatment have been successfully translated into new medications. To identify novel potential treatment targets for schizophrenia, we searched for previously unknown molecular modulators of acoustic prepulse inhibition (PPI), a schizophrenia endophenotype, in the mouse. We examined six inbred mouse strains that have a range of PPI, and used microarrays to determine which mRNA levels correlated with PPI across these mouse strains. We examined several brain regions involved in PPI and schizophrenia: hippocampus, striatum, and brainstem, found a number of transcripts that showed good correlation with PPI level, and confirmed this with real-time quantitative PCR. We then selected one candidate gene for further study, Pdxdc1 (pyridoxal-dependent decarboxylase domain containing 1), because it is a putative enzyme that could metabolize catecholamine neurotransmitters, and thus might be a feasible target for new medications. We determined that Pdxdc1 mRNA and protein are both strongly expressed in the hippocampus and levels of Pdxdc1 are inversely correlated with PPI across the six mouse strains. Using shRNA packaged in a lentiviral vector, we suppressed Pdxdc1 protein levels in the hippocampus and increased PPI by 70%. Our results suggest that Pdxdc1 may regulate PPI and could be a good target for further investigation as a potential treatment for schizophrenia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU063471 MISSION® esiRNA, esiRNA targeting mouse Pdxdc1 (esiRNA1)