EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Gardenifolins A-H, Scalemic Neolignans from Gardenia ternifolia: Chiral Resolution, Configurational Assignment, and Cytotoxic Activities against the HeLa Cancer Cell Line.
PMID 28488862
Abstract
From the tropical plant Gardenia ternifolia Schumach. and Thonn. (Rubiaceae), eight stereoisomeric 2,3-dihydrobenzo[b]furan neolignans, named gardenifolins A-H (1a-d and 2a-d), were isolated and fully structurally characterized. Reversed-phase chromatography of a stem bark extract afforded two peaks, viz. mixtures I and II, each one consisting of two diastereomers and their respective enantiomers. They were resolved and stereochemically analyzed by HPLC on a chiral phase coupled to electronic circular dichroism (ECD) spectroscopy, giving single ECD spectra of all eight stereoisomers. The double-bond geometries (E or Z) of the gardenifolins A-H and their relative configurations (cis or trans) at the stereogenic centers C-7 and C-8 in the dihydrofuran ring system were assigned by 1D and 2D NMR methods, in particular, using NOE difference experiments, whereas the absolute configurations of the isolated enantiomers were established by ECD spectroscopy by applying the reversed helicity rule. The individual pure gardenifolin isomers A-H showed the most different cytotoxic effects against the human cancer HeLa cell line, with 1d and 2a displaying the highest activities, with IC
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
577501-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
577502-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 3 cm × 2.1 mm
59352-U21 Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
59353-U21 Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm
59354-U30 Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
569422-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4 mm
569426-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
59353-U40 Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
59354-U40 Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
59352-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
569427-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
59353-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
59354-U Discovery® C8 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
59588-U Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
59590-U Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
59589-U Discovery® C8 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm