EMAIL THIS PAGE TO A FRIEND
Food chemistry
Polyunsaturated lipids and vitamin A oxidation during cod liver oil in vitro gastrointestinal digestion. Antioxidant effect of added BHT.
PMID 28490135
Abstract
The extent of cod liver oil hydrolysis and oxidation during in vitro gastrointestinal digestion was investigated by Proton Nuclear Magnetic Resonance (
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
469319
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
C8H24O2Si3