EMAIL THIS PAGE TO A FRIEND
Scientific reports
Hepatic estrogen receptor α is critical for regulation of gluconeogenesis and lipid metabolism in males.
PMID 28490809
Abstract
Impaired estrogens action is associated with features of the metabolic syndrome in animal models and humans. We sought to determine whether disruption of hepatic estrogens action in adult male mice could recapitulate aspects of the metabolic syndrome to understand the mechanistic basis for the phenotype. We found 17β-estradiol (E
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E8381
N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline sodium salt
C13H20NNaO6S