EMAIL THIS PAGE TO A FRIEND
The Journal of pathology
Cytoplasmic overexpression of RNA-binding protein HuR is a marker of poor prognosis in meningioma, and HuR knockdown decreases meningioma cell growth and resistance to hypoxia.
PMID 28493484
Abstract
HuR regulates cytoplasmic mRNA stability and translatability, and the HuR expression level has been shown to correlate with poor disease outcome in several cancer types; however, the prognostic value and potential pro-oncogenic properties of HuR in meningioma remain unclear. Thus, in the present study, we analysed 85 meningioma tissue samples to establish the relationship between HuR expression, tumour cell proliferation, and/or patient survival. In addition, we examined the anti-proliferative effects of HuR knockdown in two meningioma cell lines (IOMM-Lee and Ben-Men-1) and conducted transcriptome-wide analyses (IOMM-Lee cells) to elucidate the molecular consequences of HuR knockdown. The results of the present study showed HuR cytoplasmic expression to correlate positively with tumour grade (p = 1.2 × 10
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EHU063491 MISSION® esiRNA, esiRNA human ELAVL1 (esiRNA1)