EMAIL THIS PAGE TO A FRIEND
Research in veterinary science
Anagyrine desensitization of peripheral nicotinic acetylcholine receptors. A potential biomarker of quinolizidine alkaloid teratogenesis in cattle.
PMID 28494312
Abstract
Anagyrine, a teratogenic quinolizidine alkaloid found in Lupinus spp., is proposed to undergo metabolism by pregnant cattle to a piperidine alkaloid which inhibits fetal movement, the putative mechanism behind crooked calf syndrome. The objective of this study was to test the hypothesis that anagyrine but not lupanine or sparteine can directly, without metabolism, desensitize nicotinic acetylcholine receptors (nAChR) in a cell culture model. SH-SY5Y cells expressing autonomic nAChR, and TE-671 cells expressing fetal muscle-type nAChR were exposed to lupine alkaloids or Dimethylphenylpiperazinium (DMPP) in log