EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Superoxide-hydrogen peroxide genetic imbalance modulates differentially the oxidative metabolism on human peripheral blood mononuclear cells exposed to seleno-L-methionine.
PMID 28495429
Abstract
Superoxide-hydrogen peroxide (S-HP) imbalance genetically caused by a gene polymorphism in the human manganese superoxide dismutase enzyme (Val16Ala-MnSOD) is associated with several diseases. Into mitochondria, MnSOD catalyses superoxide radical producing HP and oxygen. Ala-MnSOD genotype presents a high MnSOD efficiency and generates the highest HP concentrations that has been associated with the risk of several cancer types. Cellular selenoenzymes glutathione peroxidase and thioredoxin reductase (TrxR) and catalase (CAT) are essential to HP removal produced in excess in cells. Since, synthesis and activities of selenoenzymes are selenium dependent, we hypothesized that AA-MnSOD cells could have an improvement on antioxidant status undergoing Seleno-L-methionine (SeMet) treatment. This study performed an in vitro protocol to evaluate the response of peripheral blood mononuclear cells (PBMC) carriers of different Val16Ala-MnSOD genotypes exposed to SeMet. SeMet effects on cell viability, apoptosis induction and modulation of oxidative variables were determined using spectrophotometric, flow cytometry, fluorimetric and immunoassays. Gene modulation of antioxidant enzymes was also performed by qRT-PCR. From an initial protocol using heterozygous (AV) cells was determined that 1nM SeMet presented a cytoprotective effect. However, whereas this concentration did not change AA viability, in VV cells it was cytotoxic by increasing necrosis events. SeMet induced higher selenoenzymes levels in AA and VV cells and decreased oxidative markers levels including DNA damage. The results suggest a pharmacogenetic positive response of SeMet effect on AA-cells. Future studies in vivo could be essential to evaluate the potential clinical impact of S-HP imbalance after use of foods or supplements containing SeMet.