EMAIL THIS PAGE TO A FRIEND
Free radical biology & medicine
Mechanism of HRP-catalyzed nitrite oxidation by H
PMID 28495446
Abstract
The peroxidative activity of horseradish peroxidase (HRP) undergoes progressive inactivation while catalyzing the oxidation of nitrite by H