EMAIL THIS PAGE TO A FRIEND
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology
Visible light initiated release of calcium ions through photochemical electron transfer reactions.
PMID 28497836
Abstract
Photolysis of anthraquinone or flavin photosensitizers in the presence of calcium EDTA complexes results in decomposition of the EDTA complex, releasing free Ca
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
743038
Sodium anthraquinone-2-sulfonate, ≥98% (HPLC)
C14H7NaO5S