EMAIL THIS PAGE TO A FRIEND
Journal of pharmacological sciences
Inhibitory effect of donepezil on bradykinin-induced increase in the intracellular calcium concentration in cultured cortical astrocytes.
PMID 28499726
Abstract
Donepezil is a potent and selective acetylcholinesterase inhibitor developed for the treatment of Alzheimer's disease. In the present study, we investigated the responses of astrocytes to bradykinin, an inflammatory mediator, and the effect of donepezil on these responses using cultured cortical astrocytes. Bradykinin induced a transient increase of intracellular calcium concentration ([Ca