Bioresource technology
Rapid biodiesel synthesis from waste pepper seeds without lipid isolation step.
PMID 28500884
In situ transformation of lipid in waste pepper seeds into biodiesel (i.e., fatty acid methyl esters: FAMEs) via thermally-induced transmethylation on silica was mainly investigated in this study. This study reported that waste pepper seeds contained 26.9wt% of lipid and that 94.1% of the total lipid in waste pepper seeds could be converted into biodiesel without lipid extraction step for only ∼1min reaction time. This study also suggested that the optimal temperature for in situ transmethylation was identified as 390°C. Moreover, comparison of in situ process via the conventional transmethylation catalyzed by H2SO4 showed that the introduced biodiesel conversion in this study had a higher tolerance against impurities, thereby being technically feasible. The in situ biodiesel production from other oil-bearing food wastes can be studied.