EMAIL THIS PAGE TO A FRIEND
Talanta
Sensitive determination of endogenous hydroxyl radical in live cell by a BODIPY based fluorescent probe.
PMID 28501175
Abstract
The sensitive and selective fluorescence probe for hydroxyl radical analysis is of significance because hydroxyl radical plays key roles in many physiological and pathological processes. In this work, a novel organic fluorescence molecular probe OHP for hydroxyl radical is synthesized by a two-step route. The probe employs 4-bora-3a,4a-diaza-s-indacene (difluoroboron dipyrromethene, BODIPY) as the fluorophore and possesses relatively high fluorescence quantum yields (77.14%). Hydroxyl radical can rapidly react with the probe and quench the fluorescence in a good linear relationship (R
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
390836
2,4-Dimethylpyrrole, 97%
C6H9N