EMAIL THIS PAGE TO A FRIEND
Talanta
A sensitive signal-off electrogenerated chemiluminescence biosensing method for the discrimination of DNA hydroxymethylation based on glycosylation modification and signal quenching from ferroceneboronic acid.
PMID 28501209
Abstract
In this study, a new and sensitive signal-off electrogenerated chemiluminescence (ECL) biosensing method for the quantification of 5-hydroxymethylcytosine in DNA (5-hmC-DNA) was developed. The method achieved simple and sensitive detection of 5-hmC-DNA based on the glycosylation of 5-hmC, combining both the amplification function of gold nanoparticles (AuNPs) and the high quenching efficiency of the tris(2, 2'-ripyridine) dichlororuthenium(II) (Ru(bpy)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
723134
Tris(2-pyridylmethyl)amine, 98%
C18H18N4