EMAIL THIS PAGE TO A FRIEND
Talanta
A modified fluorescein derivative with improved water-solubility for turn-on fluorescent determination of Hg
PMID 28501218
Abstract
To improve the water-solubility of heavy-metal sensing materials, a modified fluorescein-based derivative, acryloyl fluorescein hydrazine (ACFH), was designed and developed by incorporating a non-hydrogen-bonding group into the conjugated molecule for weakening intermolecular hydrogen-bonding interactions. In neutral water environments, ACFH presented a fluorescence-enhancement performance at λ
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
774243
1,3-Propylene sulfite, 99%
C3H6O3S