EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
The anti-asthmatic drug pranlukast suppresses the delayed-phase vomiting and reverses intracellular indices of emesis evoked by cisplatin in the least shrew (Cryptotis parva).
PMID 28501575
Abstract
The introduction of second generation serotonin 5-HT