EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The changes in glucose metabolism and cell proliferation in the kidneys of polycystic kidney disease mini-pig models.
PMID 28501615
Abstract
The pathogenic mechanism of polycystic kidney disease (PKD) is unclear. Abnormal glucose metabolism is maybe involved in hyper-proliferation of renal cyst epithelial cells. Mini-pigs are more similar to humans than rodents and therefore, are an ideal large animal model. In this study, for the first time, we systematically investigated the changes in glucose metabolism and cell proliferation signaling pathways in the kidney tissues of chronic progressive PKD mini-pig models created by knock-outing PKD1gene. The results showed that in the kidneys of PKD mini-pigs, the glycolysis is increased and the expressions of key oxidative phosphorylation enzymes Complexes I and IV significantly decreased. The activities of mitochondrial respiration chain Complexes I and IV significantly decreased; the phosphorylation level of key metabolism-modulating molecule AMP-activated protein kinase (AMPK) significantly decreased; and the mammalian target of rapamycin (mTOR) and extracellular signal-regulated kinase (ERK) signaling pathway are activated obviously. This study showed that in the kidneys of PKD mini-pigs, the level of glycolysis significantly increased, oxidative phosphorylation significantly decreased, and cell proliferation signaling pathways significantly activated, suggesting that metabolic changes in PKD may result in the occurrence and development of PKD through the activation of proliferation signaling pathways.