EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
A novel method for photo-oxidative degradation of diatrizoate in water via electromagnetic induction electrodeless lamp.
PMID 28501642
Abstract
In this study, an electromagnetic induction electrodeless lamp (EIEL) was first introduced into UV advanced oxidation processes (AOPs) for photodegradation of Diatrizoate (DTZ), which was the most persistent iodinated X-ray contrast medium (ICM), and traditional Hg lamps were taken as references. Direct photolysis rate of DTZ under EIEL irradiation was 1.34 times as that under Hg irradiation, but the electric energy consumption was 0.87 times. In this sense, the combination of EIEL and oxidants (O
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
115746
2,2,6,6-Tetramethyl-4-piperidinol, 98%
C9H19NO